Skip to Content
Merck
All Photos(1)

Key Documents

774529

Sigma-Aldrich

Titanium(IV) oxide

nanowires, diam. × L ~10 nm × 10 μm

Synonym(s):

Titania nanofibers, Titania nanowires, Titanium dioxide nanofibers, Titanium dioxide nanowires, Titanium(IV) oxide nanofibers, Titania, Titanium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

nanowires

Quality Level

diam. × L

~10 nm × 10 μm

mp

>350 °C (lit.)

density

4.26 g/mL at 25 °C (lit.)

SMILES string

O=[Ti]=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Titanium dioxide nanowires have been studied as photocatalysts[1]; battery anode materials[2]; catalyst supports[3] and as a replacement for TiO2 nanospheres in dye sensitized solar cells[4].

Preparation Note

Product prepared by hydrothermal treatment.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pan; X.;
ACS Applied Materials & Interfaces, 4, 394-3950 (2012)
Yiping; T.;
Electrochimica Acta, 78, 154-159 (2012)
Eric Formo et al.
Nano letters, 8(2), 668-672 (2008-01-22)
This paper reports a simple procedure for derivatizing the surface of anatase TiO2 nanofibers with Pt nanoparticles and then Pt nanowires. The nanofibers were prepared in the form of a nonwoven mat by electrospinning with a solution containing both poly(vinyl
Matt Law et al.
Nature materials, 4(6), 455-459 (2005-05-17)
Excitonic solar cells-including organic, hybrid organic-inorganic and dye-sensitized cells (DSCs)-are promising devices for inexpensive, large-scale solar energy conversion. The DSC is currently the most efficient and stable excitonic photocell. Central to this device is a thick nanoparticle film that provides
Nabila Haddou et al.
Chemosphere, 107, 304-310 (2014-01-28)
The Gliding Arc Discharge (GAD) is an efficient non-thermal plasma technique able to degrade organic compounds dispersed in water at atmospheric pressure. The degradation of the organometallic lead acetate (PbAc) in aqueous solution was performed by two distinct plasmageneous processes:

Articles

Dye-sensitized solar cells (DSCs) are 3rd generation solar cells combining the promise of high efficiency with low production costs.

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

See All

Questions

  1. about a carcteristic of TiO2 . it is a type N ?

    1 answer
    1. Unfortunately, there is no test on the semiconductor type of Product 774529. In general, TiO2 is intrinsically an n-type semiconductor but can be doped to have p-type semiconductivity:
      https://link.springer.com/article/10.1007/s10853-015-9303-7

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service