Skip to Content
Merck
All Photos(1)

Key Documents

771902

Sigma-Aldrich

Phenyl vinyl ether

97%

Synonym(s):

(Ethenyloxy)benzene, Phenoxyethylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O
CAS Number:
Molecular Weight:
120.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.522 (lit.)

density

0.978 g/mL at 25 °C (lit.)

functional group

phenoxy

storage temp.

2-8°C

SMILES string

C=COc1ccccc1

InChI

1S/C8H8O/c1-2-9-8-6-4-3-5-7-8/h2-7H,1H2

InChI key

NHOGGUYTANYCGQ-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.8 °F

Flash Point(C)

16 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hee-Kwon Kim et al.
ACS applied materials & interfaces, 5(12), 5648-5658 (2013-06-19)
Novel pH-responsive assemblies (PEG-lipid:DOPE liposomes) containing tunable and bifunctional phenyl-substituted vinyl ether (PIVE) cross-linkers were prepared. The assemblies consisted of 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine (DOPE), acid-cleavable poly(ethylene glycol) (PEG)-conjugated lipids, pDNA, and protamine sulfate (PS). The PIVE linkage was designed to hydrolyze under
Shihua Wang et al.
Organic letters, 13(22), 5968-5971 (2011-10-20)
Benzo[b]furans were prepared in one pot based on the addition/palladium-catalyzed C-H bond functionalization of phenols with bromoalkynes. The addition reactions of phenols to bromoalkynes generated (Z)-2-bromovinyl phenyl ethers in high yields with excellent regio- and stereoselectivity. The obtained (Z)-2-bromovinyl phenyl
M Isobe et al.
The Journal of toxicological sciences, 20(2), 161-164 (1995-05-01)
4-Nitrophenyl vinyl ether (NPVE) and phenyl vinyl ether (PVE) administered i.p. in mice lowered hepatic non-protein sulfhydryl (NP-SH) content, but did not elevate the serum glutamate pyruvate transaminase (GPT) activity. n-Butyl vinyl ether (BVE) showed no significant effects either on
Christopher L Rock et al.
Dalton transactions (Cambridge, England : 2003), 48(2), 461-467 (2018-11-30)
The phosphine-substituted α-diimine Ni precursor, (Ph2PPrDI)Ni, has been found to catalyze alkene hydrosilylation in the presence of Ph2SiH2 with turnover frequencies of up to 124 h-1 at 25 °C (990 h-1 at 60 °C). Moreover, the selective hydrosilylation of allylic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service