226475
Phenyl vinyl sulfide
97%
Synonym(s):
(Phenylthio)ethylene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5SCH=CH2
CAS Number:
Molecular Weight:
136.21
Beilstein:
969970
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.599 (lit.)
bp
94-95 °C/25 mmHg (lit.)
density
1.042 g/mL at 25 °C (lit.)
functional group
thioether
storage temp.
2-8°C
SMILES string
C=CSc1ccccc1
InChI
1S/C8H8S/c1-2-9-8-6-4-3-5-7-8/h2-7H,1H2
InChI key
GMPDOIGGGXSAPL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Phenyl vinyl sulfide was used in the synthesis of 2-fluoropyridinyl-6-oxy- precursors.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
114.8 °F - closed cup
Flash Point(C)
46 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Laurent Debien et al.
Organic letters, 15(24), 6250-6253 (2013-11-26)
2-Fluoropyridinyl-6-oxy- precursors derived from phenyl vinyl sulfide react with radicals generated from xanthates via an addition-elimination process to furnish the corresponding vinyl sulfides in good yields. This convergent method is operationally simple and enables a straightforward synthesis of the difficult
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service