Skip to Content
Merck
All Photos(1)

Key Documents

422177

Sigma-Aldrich

Ethyl vinyl ether

contains 0.1% KOH as stabilizer, 99%

Synonym(s):

Ethoxyethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH=CH2
CAS Number:
Molecular Weight:
72.11
Beilstein:
605351
EC Number:
MDL number:
UNSPSC Code:
12162002
eCl@ss:
39021025
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

liquid

contains

0.1% KOH as stabilizer
0.1% potassium hydroxide as stabilizer

refractive index

n20/D 1.376 (lit.)

bp

33 °C (lit.)

mp

−116 °C (lit.)

density

0.753 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC=C

InChI

1S/C4H8O/c1-3-5-4-2/h3H,1,4H2,2H3

InChI key

FJKIXWOMBXYWOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl vinyl ether, also known as ethoxy ethane is highly reactive due to the presence of the vinyl group and the ether functionality. It can polymerize via different methods like radical polymerization, cationic polymerization, oroxidative polymerization, yielding poly(ethyl vinyl ether) or PVE. It is widely used as a monomer to synthesize copolymers and block polymers with thermal stability and flexibility for various applications such as the synthesis of silicone hydrogel for contact lenses, tissue engineering, 3D bioprinting, drug delivery, and Li-ion batteries.

Application

Ethyl vinyl ether can be used as:


  • A monomer to synthesize amphiphilic block copolymers to fabricate protein-repelling polymersomes to produce spherical nanoparticles. These nanoparticles can be used as drug carriers.
  • A precursor to synthesize polymer electrolytes and cathode materials for solid-state lithium-ion batteries via UV photopolymerizations.
  • A monomer to synthesize poly(vinyl ether)s with controlled molecular weight and narrow dispersity via photoinduced free radical-promoted cationic reversible addition–fragmentation chain transfer (RAFT) polymerization. These polymers are used to fabricate 3D objects with different thicknesses by employing stereolithography-based 3D printing.

  • H-bonded Reusable Template Assisted para-Selective Ketonisation: This study discusses the use of ethyl vinyl ether in catalytic processes involving palladium and hexafluoroisopropanol to achieve para-selectivity in ketonisation, relevant for synthesizing complex organic compounds used in pharmaceuticals and materials science (A Maji, A Dahiya, G Lu, T Bhattacharya, 2018).

  • Mechanistic Insight into Photocontrolled Cationic Polymerization: Explores the photocontrolled polymerization of ethyl vinyl ether, providing valuable knowledge for the development of light-responsive materials, which could have applications in drug delivery and smart material systems (Q Michaudel, T Chauviré, V Kottisch, 2017).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-49.0 °F

Flash Point(C)

-45 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyglycidol-stabilized nanoparticles as a promising alternative to nanoparticle PEGylation: polymer synthesis and protein fouling considerations
Haiqin Du, et al.
Langmuir, 36, 1266-1278 (2020)
A PROXYL-Type Norbornene Polymer for High-Voltage Cathodes in Lithium Batteries
Hatakeyama-Sato, et al.
Macromolecular Rapid Communications, 42, 2100374-2100374 (2021)
Natalia Chernyak et al.
Journal of the American Chemical Society, 134(30), 12466-12469 (2012-07-21)
Anilines and ethyl vinyl ether can be used as precursors for a process that is the synthetic equivalent of the α-arylation of acetaldehyde enolate. The reaction manifests a high level of functional group compatibility, allowing the ready preparation of a
Guotao Li et al.
Journal of the American Chemical Society, 130(22), 6944-6945 (2008-05-10)
An efficient Au(I)-catalyzed synthesis of highly strained and functionalized bicyclo[3.2.0]heptanes is developed. Subsequent couplings with various nucleophiles offer additional structural features/complexity. These one-pot, three-component reactions are proposed to proceed via a key 1,3-dipolar cycloaddition between a Au carbenoid-containing carbonyl ylide
Eden Tesfu et al.
Journal of the American Chemical Society, 128(1), 70-71 (2006-01-05)
A Pd(II) reagent has been generated at preselected sites on an electrochemically addressable chip and used to effect the oxidation of the neighboring alcohols on the polymer coating the chip's surface. The resulting carbonyls were then used to accomplish site-selective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service