Skip to Content
Merck
All Photos(2)

Key Documents

768960

Sigma-Aldrich

Nitrous oxide

Messer® CANGas, 99%

Synonym(s):

Dinitrogen monoxide, Laughing gas

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
N2O
CAS Number:
Molecular Weight:
44.01
Beilstein:
8137358
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.53 (15 °C, vs air)

Quality Level

vapor pressure

51.7 mmHg ( 21 °C)

description

Filling mass: 21g

Assay

99%

form

gas

bp

−88 °C (lit.)

mp

−91 °C (lit.)

SMILES string

[O-][N+]#N

InChI

1S/N2O/c1-2-3

InChI key

GQPLMRYTRLFLPF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nitrous oxide is a gas that is colourless, odourless, and narcotic. It is also an oxidizing and liquefied gas that is heavier than air. Applications include use as the following: an oxidizing agent in atomic absorption spectrometry (AAS), oxidizer in rocket propellants, anesthetic in medical fields, propellant for whipped cream (food industries), and ultra-high purity for oxidizing and etching gas in semiconductor industries. Gas is easy to handle in a small, light pressure can, while the disposable container allows for aluminium recycling when finished.

Recommended products

Z742374: CANgas pressure regulator
Z742379: CANgas plastic hose spout
Z742378: CANgas outer adaptor
Z742377: CANgas inner adaptor
Z742375: CANgas dosing valve

Legal Information

Messer is a registered trademark of Messer Group GmbH

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Gas 1 - Press. Gas Liquefied gas - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service