769118
Xenon
99.99%, Messer® CANGas
Synonym(s):
Xenon gas
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Xe
CAS Number:
Molecular Weight:
131.29
EC Number:
MDL number:
UNSPSC Code:
12142001
PubChem Substance ID:
NACRES:
NA.22
description
Filling mass: 71g
Filling volume: 12L
Quality Level
Assay
99.99%
form
gas
SMILES string
[Xe]
InChI
1S/Xe
InChI key
FHNFHKCVQCLJFQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Xenon is a colourless and odourless rare gas that is heavier than air. Applications include use as a filling gas for lamps (halogen lamps, gas discharge lamps) and anesthetic gas in medicine. Xenon is also a component of excimer laser gas mixtures and panning gas for plasma display panels (PDP). Gas is easy to handle in a small, light pressure can, while the disposable container allows for aluminium recycling when finished.
Recommended products
Z742374: CANgas pressure regulator
Z742379: CANgas plastic hose spout
Z742378: CANgas outer adaptor
Z742377: CANgas inner adaptor
Z742375: CANgas dosing valve
Z742379: CANgas plastic hose spout
Z742378: CANgas outer adaptor
Z742377: CANgas inner adaptor
Z742375: CANgas dosing valve
Legal Information
Messer is a registered trademark of Messer Group GmbH
accessory
Product No.
Description
Pricing
regulator
Product No.
Description
Pricing
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Press. Gas Liquefied gas
Storage Class Code
2A - Gases
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service