Ethylene - 768987 is a 99.95% pure gas that is not diluted in anything.
768987
Ethylene
Messer® CANGas, 99.95%
Synonym(s):
Ethene
About This Item
Recommended Products
vapor density
0.97 (vs air)
Quality Level
vapor pressure
35.04 atm ( 20 °C)
description
Filling mass: 15g
Filling volume: 12L
Assay
99.95%
form
gas
autoignition temp.
842 °F
expl. lim.
36 %
bp
−104 °C (lit.)
mp
−169 °C (lit.)
SMILES string
C=C
InChI
1S/C2H4/c1-2/h1-2H2
InChI key
VGGSQFUCUMXWEO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Recommended products
Z742379: CANgas plastic hose spout
Z742378: CANgas outer adaptor
Z742377: CANgas inner adaptor
Z742375: CANgas dosing valve
Legal Information
accessory
regulator
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Gas 1 - Press. Gas Liquefied gas - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
2A - Gases
WGK
nwg
Flash Point(F)
-148.0 °F - closed cup
Flash Point(C)
-100 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Hi there, I want to use this gas (ethylene) in UHV for adsorption. I would like to confirm that the bottle contains only (99.995%) ethylene, so I will not suffer from other admixtures. In general, is ethylene diluted in air or some other gases?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service