Skip to Content
Merck
All Photos(3)

Key Documents

762377

Sigma-Aldrich

Sodium trifluoromethanesulfonimide

97%

Synonym(s):

Sodium bis(trifluoromethylsulfonyl)imide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2F6NNaO4S2
CAS Number:
Molecular Weight:
303.14
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

257.1-258.4 °C

SMILES string

FC(F)(F)S(=O)(=O)N([Na])S(=O)(=O)C(F)(F)F

InChI

1S/C2F6NO4S2.Na/c3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;/q-1;+1

InChI key

YLKTWKVVQDCJFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A sodiumbis(trifluoromethylsulfonyl)imide serves as an electrolyte in batteries and fuel cells. Additionally, it is used as a Lewis acid and ionic liquid.

Application

Uses:
  • Electrolyte for batteries and fuel cells
  • Ionic liquids
  • Lewis acids

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple access to metallic or onium bistrifluoromethanesulfonimide salts
Arvai R, et al.
Tetrahedron, 65(27), 5361-5368 (2009)
A simple access to metallic or onium bistrifluoromethanesulfonimide salts
Arvai, R.; et al.
Tetrahedron, 65, 5361-5368 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service