Skip to Content
Merck
All Photos(2)

Key Documents

753424

Sigma-Aldrich

Magnesium bis(trifluoromethanesulfonimide)

Synonym(s):

Magnesium bis(ditriflimide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4F12MgN2O8S4
CAS Number:
Molecular Weight:
584.60
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

mp

>200 °C

SMILES string

FC(F)(F)S(=O)(=O)N([Mg]N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

InChI

1S/2C2F6NO4S2.Mg/c2*3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;/q2*-1;+2

InChI key

DMFBPGIDUUNBRU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium bis(trifluoromethanesulfonimide) (Mg(TFSI)2) is a strong magnesium Lewis acid. It is used as a catalyst in organic synthesis. For instance, it is used in the synthesis of dihydropyrazoles by reaction of nitrilimines with alkenes. Additionally, Mg(TFSI)2 salt is also useful for ionic conductivity studies, transference number measurements, and electrochemical properties of gel polymer electrolyte (GPE) systems.
Magnesium bis(trifluoromethanesulfonimide), also known as magnesium triflimide, can be used as an inorganic catalyst in various organic reactions, including acetylation of phenols and alcohols, aminolysis of lactones with amines, [2 + 2] cycloadditions of siloxy-alkynes with carbonyl compounds, cycloisomerization of 1,6-dienes, Friedel-Crafts acylation, and for the synthesis of coumarins.

Application

  • Rechargeable Battery Research: Utilized in the study of rechargeable Mg-ion batteries, Magnesium bis(trifluoromethanesulfonimide) serves as a model anode material, providing significant insights into atomistic and mesoscale mechanisms that could revolutionize battery development and application (Kravchyk et al., 2018).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnesium bis (trifluoromethane) sulfonimide: An efficient catalyst for the synthesis of coumarins under solvent-free conditions
Wang H
Monatshefte fur Chemie / Chemical Monthly, 144, 411-414 (2013)
Magnesium Bis (trifluoromethylsulfonyl) imide
Selvakumar S, et al.
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Toshihiko Mandai et al.
Physical chemistry chemical physics : PCCP, 21(23), 12100-12111 (2019-04-26)
To achieve a sustainable-energy society in the future, next-generation highly efficient energy storage technologies, particularly those based on multivalent metal negative electrodes, are urgently required to be developed. Magnesium rechargeable batteries (MRBs) are promising options owing to the many advantageous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service