Skip to Content
Merck
All Photos(1)

Key Documents

464635

Sigma-Aldrich

Trifluoromethanesulfonimide

≥95.0% (19F-NMR)

Synonym(s):

Bis(trifluoromethanesulfonyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO2)2NH
Molecular Weight:
281.15
Beilstein:
4754101
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (19F-NMR)

form

solid

bp

90-91 °C (lit.)

mp

46-57 °C (lit.)

functional group

fluoro

SMILES string

FC(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F

InChI

1S/C2HF6NO4S2/c3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h9H

InChI key

ZXMGHDIOOHOAAE-UHFFFAOYSA-N

General description

Trifluoromethanesulfonimide is a promising Bronsted acid catalyst and is superior to scandium triflate in catalytic action.

Application

Trifluoromethanesulfonimide may be used as a catalyst for the preparation of 1-substituted-1H-1,2,3,4-tetrazoles.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trifluoromethanesulfonimide catalysed synthesis of 1-substituted-1H-1, 2, 3, 4-tetrazoles using glycerol as green solvent at room temperature.
Wang H, et al.
J. Chem. Res. (M), 40(9), 570-572 (2016)
Practical Synthesis of (?)-a-Tocopherol. Trifluoromethanesulfonimide as an Extremely Active Br?nsted Acid Catalyst for the Condensation of Trimethylhydroquinone with Isophytol.
Ishihara K, et al.
Synlett, 11, 1045-1046 (1996)
Youngbum Kim et al.
Nanoscale, 10(18), 8851-8858 (2018-05-02)
The exciton-dominated light emission of two-dimensional (2D) semiconductors is determined largely by the doping state and the formation of defects. Extensive studies have shown that chemical treatment critically modifies the doping state and defect state of chemical vapor deposition (CVD)-grown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service