496952
2-[[(Butylamino)carbonyl]oxy]ethyl acrylate
Synonym(s):
2-(N -Butylcarbamoyloxy)ethyl acrylate, 2-[[(Butylamino)carbonyl]oxy]ethyl acrylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
H2C=CHCO2CH2CH2O2CNH(CH2)3CH3
CAS Number:
Molecular Weight:
215.25
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
refractive index
n20/D 1.46 (lit.)
density
1.06 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CCCCNC(=O)OCCOC(=O)C=C
InChI
1S/C10H17NO4/c1-3-5-6-11-10(13)15-8-7-14-9(12)4-2/h4H,2-3,5-8H2,1H3,(H,11,13)
InChI key
FLKHVLRENDBIDB-UHFFFAOYSA-N
Application
- Well-Tunable, 3D-printable, and Fast Autonomous Self-Healing Elastomers: This study discusses the use of 2-[[(Butylamino)carbonyl]oxy]ethyl acrylate monomer, which was purified to remove inhibitors through a basic Al2O3 column. The monomer′s properties contribute to developing self-healing elastomers (B Li, S Ge, X Zhao, Q Chen, J Tian, D Hun, 2023 - Elsevier). Read more.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service