Skip to Content
Merck
All Photos(1)

Key Documents

407283

Sigma-Aldrich

Dipentaerythritol penta-/hexa-acrylate

contains ≤650 ppm MEHQ as inhibitor

Synonym(s):

Dipentaerythritol monohydroxypentaacrylate, Dipentaerythritol pentaacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O[CH2C(CH2OR)3]2, R = COCH=CH2 or H
CAS Number:
Molecular Weight:
524.51
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

Quality Level

contains

≤650 ppm MEHQ as inhibitor

refractive index

n20/D 1.49 (lit.)

density

1.155 g/mL at 25 °C (lit.)

SMILES string

OCC(COCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C.C=CC(=O)OCC(COCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C

InChI

1S/C28H34O13.C25H32O12/c1-7-21(29)36-15-27(16-37-22(30)8-2,17-38-23(31)9-3)13-35-14-28(18-39-24(32)10-4,19-40-25(33)11-5)20-41-26(34)12-6;1-6-19(27)33-14-24(11-26,15-34-20(28)7-2)12-32-13-25(16-35-21(29)8-3,17-36-22(30)9-4)18-37-23(31)10-5/h7-12H,1-6,13-20H2;6-10,26H,1-5,11-18H2

InChI key

WDCLDIHFVMMPSW-UHFFFAOYSA-N

General description

Dipentaerythritol penta-/hexa-acrylate (DPHPA) is a multifunctional acrylate.

Application

DPHPA may be used to prepare nanoporous/non porous polymeric thin films for drug delivery systems. One of the constituents of prepolymer syrup to fabricate holographic polymer-dispersed liquid crystals(H-PDLCs) gratings is DPHPA . It may be used to form a photopolymer blend for various applications such as microelectronics and micro-optical elements in displays.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photoembossing of Periodic Relief Structures Using Polymerization?Induced Diffusion: A Combinatorial Study.
Sanchez C, et al.
Advanced Materials, 17(21), 2567-2571 (2005)
Optically switchable gratings based on azo-dye-doped, polymer-dispersed liquid crystals.
Liu YJ, et al.
Optics Letters, 34(15), 2351-2353 (2009)
Towards nanoporous polymer thin film-based drug delivery systems.
Yan W, et al.
Thin Solid Films, 517(5), 1794-1798 (2009)
All-optical modulation of localized surface plasmon coupling in a hybrid system composed of photoswitchable gratings and Au nanodisk arrays.
Liu YJ, et al.
The Journal of Physical Chemistry C, 115(15), 7717-7722 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service