Skip to Content
Merck
All Photos(1)

Key Documents

744867

Sigma-Aldrich

(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyl N-succinimidyl carbonate

for Copper-free Click Chemistry

Synonym(s):

N-[((1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-yl)methyloxycarbonyloxy]succinimide, BCN-NHS, BCN-succinimidyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17NO5
CAS Number:
Molecular Weight:
291.30
UNSPSC Code:
12161502
PubChem Substance ID:
NACRES:
NA.22

Quality Level

form

powder

composition

carbon content, 61.85%
hydrogen content, 5.88%
nitrogen content, 4.81%

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

functional group

NHS ester

storage temp.

−20°C

SMILES string

[H][C@@]12CCC#CCC[C@]1([H])[C@@H]2COC(ON3C(CCC3=O)=O)=O

InChI

1S/C15H17NO5/c17-13-7-8-14(18)16(13)21-15(19)20-9-12-10-5-3-1-2-4-6-11(10)12/h10-12H,3-9H2/t10-,11+,12-

InChI key

SKTDJYHCSCYLQU-ZSBIGDGJSA-N

Application

(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyl N-succinimidyl carbonate may be used to functionalize the primary amines of the lysine residue of recombinant elastin-like proteins (ELPs). These functionalized ELPs quickly undergo cross-linking via bio-orthogonal strain-promoted azide-alkyne cycloaddition reactions to form hydrogels with high gelation kinetics and tunable mechanics, making them useful biomaterials for cell encapsulation.{67
It may also be used to synthesize bicyclononyne functionalized poly(ethylene glycol) polymer coatings with anti-fouling properties towards protein adhesion and cell adhesion for supramolecular ureidopyrimidinone (UPy) based materials.
Succinimidyl ester/NHS functionalized cyclooctyne derivative for incorporation of the cyclooctyne moiety into amine containing compounds or biomolecules. Cyclooctynes are useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. This strained cyclooctyne will react with azide functionalized compounds or biomolecules without the need for a copper catalyst to result in a stable triazole linkage.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

?Click? access to multilayer functionalized Au surface: A terpyridine patterning example.
Materials Science and Engineering, C, 75, 1343-1350 (2017)
Introduction of anti-fouling coatings at the surface of supramolecular elastomeric materials via post-modification of reactive supramolecular additives
Goor OJ, et al.
Polym. Chem., 8(34), 5228-5238 (2017)
Installing an additional emission quenching pathway in the design of iridium (III)-based phosphorogenic biomaterials for bioorthogonal labelling and imaging
Li SPY, et al.
Biomaterials, 103, 305-313 (2016)
Structural Manipulation of Ruthenium (II) Polypyridine Nitrone Complexes to Generate Phosphorogenic Bioorthogonal Reagents for Selective Cellular Labeling
Tang TSM, et al.
Chemistry?A European Journal , 22(28), 9649-9659 (2016)
Recombinant DNA technology and click chemistry: a powerful combination for generating a hybrid elastin-like-statherin hydrogel to control calcium phosphate mineralization.
Misbah MH, et al.
Beilstein Journal of Nanotechnology, 8, 772-772 (2017)

Articles

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service