Skip to Content
Merck
All Photos(1)

Key Documents

715166

Sigma-Aldrich

Octadecylphosphonic acid

97%

Synonym(s):

n-Octadecylphosphonic acid, ODPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H39O3P
CAS Number:
Molecular Weight:
334.47
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

crystals

mp

95-100 °C

SMILES string

CCCCCCCCCCCCCCCCCCP(O)(O)=O

InChI

1S/C18H39O3P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-22(19,20)21/h2-18H2,1H3,(H2,19,20,21)

InChI key

FTMKAMVLFVRZQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octadecylphosphonic acid (OPA) is used as a self-assembled monolayer that forms a surfactant which strongly bonds with the surface atoms of different metal oxides. It provides hydrophobic properties and covers a higher surface area than the predominantly used silanes.

Application

Anti-corrosive coatings on cupronickel (CuNi) can be formed by fabricating a protective film of OPA by dip coating method. OPA may be used in the preparation of indium oxide(In2O3) nanowire transistor than can find potential application in the growth of fast switching and low noise nano-electronic devices. It may be used for self-cleaning and oil/water separation based applications as it forms a superhydrophobic layer on the copper mesh.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Getachew Tizazu et al.
Langmuir : the ACS journal of surfaces and colloids, 25(18), 10746-10753 (2009-07-18)
Electron-hole pair formation at titania surfaces leads to the formation of reactive species that degrade organic materials. Here, we describe the degradation of self-assembled monolayers of alkylphosphonic acids on the native oxide of titanium following exposure to UV light. The
Fabrication of controllable and stable In2O3 nanowire transistors using an octadecylphosphonic acid self-assembled monolayer.
Lim T, et al.
Nanotechnology, 26(14), 145203-145203 (2015)
Resistance of Superhydrophobic Surface-Functionalized TiO2 Nanotubes to Corrosion and Intense Cavitation.
Hua W.
Nanomaterials, 8(10), 783-783 (2018)
Fast formation of superhydrophobic octadecylphosphonic acid (ODPA) coating for self-cleaning and oil/water separation.
Dai C, et al.
Soft Matter, 10(40), 8116-8121 (2014)
n-Octadecylphosphonic acid grafted mesoporous magnetic nanoparticle: Preparation, characterization, and application in magnetic solid-phase extraction.
Ding J, et al.
Journal of Chromatography A, 1217(47), 7351-7358 (2010)

Articles

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service