759902
Dibenzocyclooctyne-PEG4-acid
for Copper-free Click Chemistry
Synonym(s):
Polyethylene glycol, DBCO-PEG4-Acid
About This Item
Recommended Products
form
solid
reaction suitability
reaction type: click chemistry
reagent type: linker
functional group
carboxylic acid
storage temp.
−20°C
SMILES string
O=C(CCCCC(NCCOCCOCCOCCOCCC(O)=O)=O)N1CC2=C(C=CC=C2)C#CC3=C1C=CC=C3
InChI
1S/C32H40N2O8/c35-30(33-16-18-40-20-22-42-24-23-41-21-19-39-17-15-32(37)38)11-5-6-12-31(36)34-25-28-9-2-1-7-26(28)13-14-27-8-3-4-10-29(27)34/h1-4,7-10H,5-6,11-12,15-25H2,(H,33,35)(H,37,38)
InChI key
RMYANOWYMFCGGS-UHFFFAOYSA-N
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.
Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.
Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.
Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service