Skip to Content
Merck
All Photos(1)

Key Documents

743062

Sigma-Aldrich

TurboBeads Complexon

extent of labeling: ≥0.1 mmol/g loading (-Ph-CH2-N(CH2-COOH)2)

Synonym(s):

Nano particles, magnetic, IDA functionalized

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352128
NACRES:
NA.22

product line

TurboBeads

Quality Level

Assay

≥99%

form

powder

composition

carbon content, ≤14 wt. %

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: copper reactive

extent of labeling

≥0.1 mmol/g loading (-Ph-CH2-N(CH2-COOH)2)

magnetization

≥120 emu/g, mass saturation

surface area

≥15 m2/g

average diameter

≤50 nm

suitability

conforms to structure for Infrared spectrum

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

Proof of identity: metallic cobalt identity from XRD

air-stability:
weight gain in air at 400°C >20 wt.%
weight gain in air at 100°C <3 wt.%

Legal Information

TurboBeads is a trademark of TurboBeads LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quirin M Kainz et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(30), 10038-10045 (2013-06-19)
Unprecedented magnetic borohydride exchange (mBER), magnetic Wang aldehyde (mWang) and magnetic amine resins were prepared from highly magnetic polymer-coated cobalt or iron nanoparticles. Microwave irradiation was used to obtain excellent degrees of functionalization (>95 %) and loadings (up to 3.0 mmol  g(-1))
Jacinto Sá et al.
Nanoscale, 5(18), 8462-8465 (2013-07-12)
Non-magnetic Pt catalysts, supported on carbon coated magnetic Co nanoparticles, changed catalytic performance in the presence of an external magnetic field. This behavior relates to an electronic change of Pt induced by a localized magnetic field, which modifies the CO
Vladimir Zlateski et al.
Bioconjugate chemistry, 25(4), 677-684 (2014-03-29)
In the pursuit of robust and reusable biocatalysts for industrial synthetic chemistry, nanobiotechnology is currently taking a significant part. Recently, enzymes have been immobilized on different nanoscaffold supports. Carbon coated metallic nanoparticles were found to be a practically useful support
Inge K Herrmann et al.
Nanoscale, 5(18), 8718-8723 (2013-08-01)
In a number of clinical conditions such as intoxication, bacteraemia or autoimmune diseases the removal of the disease-causing factor from blood would be the most direct cure. However, physicochemical characteristics of the target compounds limit the applicability of classical filtration
Saroj Kumar et al.
Journal of nanobiotechnology, 11, 14-14 (2013-05-04)
Introduction of effective point-of-care devices for use in medical diagnostics is part of strategies to combat accelerating health-care costs. Molecular motor driven nanodevices have unique potentials in this regard due to unprecedented level of miniaturization and independence of external pumps.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service