Skip to Content
Merck
All Photos(1)

Key Documents

900957

Sigma-Aldrich

Azide agarose

Synonym(s):

Azide functionalized agarose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352125
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reaction type: click chemistry

refractive index

n/D 1.343

density

0.978 g/mL

storage temp.

2-8°C

Application

Azide agarose is a 6% crosslinked agarose resin that is activated with azide functional groups for covalent capture of alkyne-tagged biomolecules via copper(I)-catalyzed alkyne-azide or copper(I)-free strain-promoted alkyne-azide click chemistry approach. This product can be used for either a batch protocol or a low-pressure column method.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

?Clickable? agarose for affinity chromatography.
Punna S, et al.
Bioconjugate Chemistry, 16(6), 1536-1541 (2005)
Sreenivas Punna et al.
Bioconjugate chemistry, 16(6), 1536-1541 (2005-11-17)
Successful purification of biological molecules by affinity chromatography requires the attachment of desired ligands to biocompatible chromatographic supports. The Cu(I)-catalyzed cycloaddition of azides and alkynes-the premier example of "click chemistry"-is an efficient way to make covalent connections among diverse molecules

Articles

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service