Skip to Content
Merck
All Photos(1)

Key Documents

GE45152105050250

SpeedBeads magnetic carboxylate modified particles

Cytiva 45152105050250, 1 μm avg. part. size, suspension (5% Solids)

Synonym(s):

SpeedBeads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.56

ligand

(carboxylate modified particles)

form

suspension (5% Solids)

packaging

pkg of 15 mL

manufacturer/tradename

Cytiva 45152105050250

concentration

50 mg/mL

avg. part. size

1 μm

shipped in

wet ice

storage temp.

2-8°C

Related Categories

Application

Sera-Mag SpeedBeads and Sera-Mag Carboxylate-Modified Magnetic Particles typical application uses include sample preparation, proteomics, nucleic acid isolation, and immunoassay applications.

Features and Benefits

Sera-Mag SpeedBeads and Sera-Mag Carboxylate-Modified Magnetic Particles allow convenient covalent coupling of target molecules.

  • Carboxylic groups on the surface permit easy covalent coupling to biomolecules of interest using convenient carbodiimide chemistry.
  • Different levels of hydrophobicity/hydrophilicity available.
  • Available in Sera-Mag SpeedBeads and original Sera-Mag versions.
  • Excellent sensitivity and low nonspecific binding for greater accuracy.
  • Fast Magnetic Response Times and Stable Physical Integrity

Storage and Stability

4 °C

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sera-Mag is a trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magdalena Surman et al.
International journal of molecular sciences, 21(8) (2020-04-26)
Cutaneous melanoma (CM) is an aggressive type of skin cancer for which effective biomarkers are still needed. Recently, the protein content of extracellular vesicles (ectosomes and exosomes) became increasingly investigated in terms of its functional role in CM and as
Jan Leipert et al.
Lab on a chip, 19(20), 3490-3498 (2019-09-19)
While LC-MS-based proteomics with high nanograms to micrograms of total protein has become routine, the analysis of samples derived from low cell numbers is challenged by factors such as sample losses, or difficulties encountered with the manual manipulation of small
Georg Michlits et al.
Nature methods, 14(12), 1191-1197 (2017-10-19)
Pooled CRISPR screens are a powerful tool for assessments of gene function. However, conventional analysis is based exclusively on the relative abundance of integrated single guide RNAs (sgRNAs) between populations, which does not discern distinct phenotypes and editing outcomes generated

Protocols

Covalent coupling of antibodies, aptamers, and proteins with a NHS Mag Sepharose, Sera-Mag Carboxylate-Modified, Sera-Mag SpeedBeads Carboxylate-Modified from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service