Skip to Content
Merck
All Photos(1)

Key Documents

73815

Sigma-Aldrich

3-Nitrosalicylic acid

≥98.0% (HPLC)

Synonym(s):

2-Hydroxy-3-nitrobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO5
CAS Number:
Molecular Weight:
183.12
Beilstein:
2213132
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (HPLC)

form

crystals

mp

142-147 °C

functional group

carboxylic acid

SMILES string

OC(=O)c1cccc(c1O)[N+]([O-])=O

InChI

1S/C7H5NO5/c9-6-4(7(10)11)2-1-3-5(6)8(12)13/h1-3,9H,(H,10,11)

InChI key

WWWFHFGUOIQNJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Nitrosalicylic acid (3NSA) is generally used as a corrosion inhibitor. It can be used as a supporting electrolyte for the electrodeposition of pyrrole on zinc for corrosion protection. 3NSA can also be used as a ligand for the synthesis of complexes with iron and aluminum for chelating therapy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of substituents on complex stability aimed at designing new iron (III) and aluminum (III) chelators.
Nurchi VM, et al.
Journal of Inorganic Biochemistry, 103(2), 227-236 (2009)
Release mechanism of electrodeposited polypyrrole doped with corrosion inhibitor anions.
Paliwoda-Porebska G, et al.
Journal of Solid State Electrochemistry, 10(9), 730-736 (2006)
Stimuli-selective delivery of two payloads from dual responsive nanocontainers.
Lv LP, et al.
Chemistry of Materials, 26(11), 3351-3353 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service