3-Nitrosalicylic acid
Synonym(s):
2-Hydroxy-3-nitrobenzoic acid
Empirical Formula (Hill Notation):
C7H5NO5
CAS Number:
Molecular Weight:
183.12
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2