Skip to Content
Merck
All Photos(1)

Key Documents

675725

Sigma-Aldrich

4-Bromo-1-butyne

97%

Synonym(s):

1-Bromo-3-butyne

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5Br
CAS Number:
Molecular Weight:
132.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.481

density

1.417 g/mL at 25 °C

SMILES string

BrCCC#C

InChI

1S/C4H5Br/c1-2-3-4-5/h1H,3-4H2

InChI key

XLYOGWXIKVUXCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Bromo-1-butyne is commonly used as a reactant. It serves as a source of alkyl halides for the introduction of bromo functionality into the molecule.

Application

4-Bromo-1-butyne is used as a reactant in the synthesis of:
  • Macrocycles by cobalt-mediated [2+2+2] co-cyclotrimerization.
  • 2,4,5-trisubstituted oxazoles by a gold-catalyzed formal [3+2] cycloaddition.
  • Intramolecular 1,3-dipolar cycloaddition to synthesize 1,3,4-oxadiazoles.
  • Lactones bearing alkynes for reductive cyclization in the preparation of azulene derivatives.
  • Substituted α-pyrones by gold-catalyzed coupling reactions.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.0 °F

Flash Point(C)

23.9 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intermolecular and Selective Synthesis of 2, 4, 5-Trisubstituted Oxazoles by a Gold-Catalyzed Formal [3+ 2] Cycloaddition.
Davies PW, et al.
Angewandte Chemie (International Edition in English), 50(38), 8931-8935 (2011)
Reductive Cyclization Cascades of Lactones Using SmI2- H2O.
Parmar D, et al.
Journal of the American Chemical Society, 133(8), 2418-2420 (2011)
Synthesis of macrocycles via cobalt-mediated [2+ 2+ 2] cycloadditions.
Bonaga LVR, et al.
Journal of the American Chemical Society, 127(10), 3473-3485 (2005)
A facile synthesis of N-C linked 1, 2, 3-triazole-oligomers
V Fiandanese, et al.
Tetrahedron, 67, 5254-5260 (2011)
Intramolecular Diels- Alder/1, 3-dipolar cycloaddition cascade of 1, 3, 4-oxadiazoles.
Elliott GI, et al.
Journal of the American Chemical Society, 128(32), 10589-10595 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service