711357
4-Aminotetrahydropyran
97%
Synonym(s):
Tetrahydro-2H-pyran-4-amine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.463
density
0.977 g/cm3 at 25 °C
functional group
ether
SMILES string
NC1CCOCC1
InChI
1S/C5H11NO/c6-5-1-3-7-4-2-5/h5H,1-4,6H2
InChI key
AHVQYHFYQWKUKB-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
129.9 °F - closed cup
Flash Point(C)
54.4 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Physical chemistry chemical physics : PCCP, 22(7), 4086-4095 (2020-02-08)
Light-responsive binary (azobenzene + solvent) lyotropic liquid crystals (LCs) were investigated by structural modification of simple azobenzene molecules. Three benzoic acid-containing azobenzene molecules 4-(4-(hydroxyphenyl)diazenyl)benzoic acid (AZO1), 3-(4-(hydroxyphenyl)diazenyl)benzoic acid (AZO2) and 5-(4-(hydroxyphenyl)diazenyl)isophthalic acid (AZO3) were produced with various amide substitutions to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service