Skip to Content
Merck
All Photos(1)

Key Documents

710784

Sigma-Aldrich

11-Deoxycortisol-2,2,4,6,6-d5

98 atom % D, 98% (CP)

Synonym(s):

11-Deoxy-17-hydroxycorticosterone-2,2,4,6,6-d5, 17,21-Dihydroxyprogesterone-2,2,4,6,6-d5, Cortexolone-2,2,4,6,6-d5, Reichstein′s substance S (2,2,4,6,6-d5)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21D5H25O4
CAS Number:
Molecular Weight:
351.49
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

98 atom % D

Quality Level

Assay

98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

208 °C (dec.)

mass shift

M+5

storage temp.

−20°C

SMILES string

[2H]C1=C2C([2H])([2H])C[C@@H]3[C@H](CC[C@@]4(C)[C@H]3CC[C@]4(O)C(=O)CO)[C@@]2(C)CC([2H])([2H])C1=O

InChI

1S/C21H30O4/c1-19-8-5-14(23)11-13(19)3-4-15-16(19)6-9-20(2)17(15)7-10-21(20,25)18(24)12-22/h11,15-17,22,25H,3-10,12H2,1-2H3/t15-,16+,17+,19+,20+,21+/m1/s1/i3D2,5D2,11D

InChI key

WHBHBVVOGNECLV-NGKMQDFHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silke Matysik et al.
Journal of chromatography. A, 1526, 112-118 (2017-10-25)
A limited specificity is inherent to immunoassays for steroid hormone analysis. To improve selectivity mass spectrometric analysis of steroid hormones by liquid chromatography-tandem mass spectrometry (LC-MS/MS) has been introduced in the clinical laboratory over the past years usually with low
Teng-Fei Yuan et al.
Journal of lipid research, 61(4), 580-586 (2020-01-23)
Analyzing global steroid metabolism in humans can shed light on the etiologies of steroid-related diseases. However, existing methods require large amounts of serum and lack the evaluation of accuracy. Here, we developed an LC/MS/MS method for the simultaneous quantification of
Mirko Peitzsch et al.
The Journal of steroid biochemistry and molecular biology, 145, 75-84 (2014-10-15)
Steroid profiling for diagnosis of endocrine disorders featuring disordered production of steroid hormones is now possible from advances in liquid chromatography with tandem mass spectrometry (LC-MS/MS). Adrenal venous (AV) measurements of aldosterone and cortisol are a standard practice in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service