Skip to Content
Merck
All Photos(1)

Key Documents

D-076

Supelco

21-Deoxycortisol-d8 (2,2,4,6,6,21,21,21-d8)

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

21-Deoxycortisol-D8 (2,2,4,6,6,21,21,21-D8) solution, 17α-Dihydroxyprogesterone-D8 (2,2,4,6,6,21,21,21-D8) solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22O4D8
Molecular Weight:
354.51
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C1=C2C([2H])([2H])C[C@H]3[C@@H]4CC[C@](O)(C(=O)C([2H])([2H])[2H])[C@@]4(C)C[C@H](O)[C@@H]3[C@@]2(C)CC([2H])([2H])C1=O

InChI

1S/C21H30O4/c1-12(22)21(25)9-7-16-15-5-4-13-10-14(23)6-8-19(13,2)18(15)17(24)11-20(16,21)3/h10,15-18,24-25H,4-9,11H2,1-3H3/t15-,16-,17-,18+,19-,20-,21-/m0/s1/i1D3,4D2,6D2,10D

InChI key

LCZBQMKVFQNSJR-ZERYXUNYSA-N

General description

21-Deoxycortisol is a steroid intermediate in the formation of cortisol, an important glucocorticoid in pregnancy and fetal development. This stable labeled internal standard is suitable for quantitation of 21-Deoxycortisol levels in LC-MS/MS testing assays for newborn screening or endocrinology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service