Skip to Content
Merck
All Photos(1)

Key Documents

C-113

Supelco

Cortisol-D4 (9,11,12, 12-D4) solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Cortisol-D4 (9,11,12,12-D4) solution, Hydrocortisone-9,11,12,12-D4 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26D4O5
CAS Number:
Molecular Weight:
366.48
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C1([2H])[C@]([2H])(O)[C@@]2([2H])[C@@H](CCC3=CC(=O)CC[C@]23C)[C@@H]4CCC(O)(C(=O)CO)[C@@]14C

InChI

1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1/i10D2,16D,18D

InChI key

JYGXADMDTFJGBT-BJJKTIQHSA-N

General description

Cortisol-D4 (9,11,12, 12-D4) is the deuterated form of the glucocorticoid, cortisol. This stable labeled internal standard is suitable for quantitation of cortisol levels in whole blood by LC-MS/MS for newborn screening of congenital adrenal hyperplasia (CAH) and in serum or plasma by LC-MS/MS for determination of several diseases including Cushing’s syndrome, Addison’s disease, and Sheehan’s syndrome.

Application

This Certified Spiking Solution® is suitable for GC/MS and LC/MS applications from clinical and diagnostic testing and endocrinology to sports testing.The Certified Reference Material (CRM) in solution can also be used as an internal standard as follows:
  • Identification of cortisol by atmospheric pressure chemical ionization (APCI) and selected reaction monitoring (SRM) during the high-performance liquid chromatography- tandem mass spectrometric (HPLC-MS/MS) analysis of cortisol in human sweat samples
  • Combined determination of cortisol and cortisone in 48 serum samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • Development and validation of an LC-MS/MS method to determine five steroids― free cortisol, cortisone, tetrahydrocortisol, allo-tetrahydrocortisol, and tetrahydrocortisone in urine samples
  • Multi-residue analysis of cortisol, cortisone, and dehydroepiandrosterone sulfate (DHEAS) in 103 hair samples by liquid chromatography-electrospray ionization-tandem mass spectrometry (LC-MS/MS)
  • Rapid LC-MS/MS-based assay of cortisol in 2860 serum samples over a duration of one year, using a simple protein precipitation technique for sample preparation

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan P Sanabria-Mazo et al.
BMJ open, 10(7), e038107-e038107 (2020-07-28)
The IMPACT study focuses on chronic low back pain (CLBP) and depression symptoms, a prevalent and complex problem that represents a challenge for health professionals. Acceptance and Commitment Therapy (ACT) and Brief Behavioural Activation Treatment for Depression (BATD) are effective
Helen V Ratajczak et al.
Future science OA, 1(4), FSO70-FSO70 (2016-12-30)
Measure biomarkers pertinent to autism in saliva from humans. At 7:30 PM (reading instructions) and 8:30 PM (hearing instructions), neurotypical adults (6 M, 6 F) each spat into tubes containing protease inhibitors. Cells were counted, samples aliquoted, frozen and thawed.
X Yang et al.
Cell death & disease, 5, e1308-e1308 (2014-06-27)
Chronic psychological stress has been demonstrated to play an important role in several severe diseases, but whether it affects disease therapy or not remains unclear. Mesenchymal stem cells (MSCs) have been demonstrated to have therapeutic potentials in treating tissue injury

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service