Skip to Content
Merck
All Photos(2)

Key Documents

700045

Sigma-Aldrich

(2-Butenyl)chloropalladium dimer

≥97%

Synonym(s):

Bis[(1,2,3-η)-2-buten-1-yl]di-μ-chlorodi-palladiumdi-π-crotylpalladium chloride, Di-π-Crotylpalladium chloride, Di-2-butenyldipalladium dichloride, Dichlorobis(1-methylallyl)dipalladium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14Cl2Pd2
CAS Number:
Molecular Weight:
393.94
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

133-140 °C

SMILES string

C\C=C\C[Pd]Cl.C\C=C\C[Pd]Cl

InChI

1S/2C4H7.2ClH.2Pd/c2*1-3-4-2;;;;/h2*3-4H,1H2,2H3;2*1H;;/q;;;;2*+1/p-2/b2*4-3+;;;;

InChI key

NTFUUQYNUUTALG-HYTOEPEZSA-L

Application

Catalyst for:
  • Enantioselective elimination of palladium hydride
  • Non-covalent anchoring of catalysts to silica supports
  • Allylations of cyanoacetates
  • Enantioselective nucleophilic substitutions

Reactant for:
  • Allylic alkylation reactions
  • Preparation of phosphane-functionalized carbosilane dendrimers

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service