Skip to Content
Merck
All Photos(6)

Key Documents

222380

Sigma-Aldrich

Allylpalladium(II) chloride dimer

greener alternative

98%

Synonym(s):

Bis((η3-allyl)(chloro)palladium), Di-ο-allyldi-μ-chlorodipalladium, [PdCl(C3H5)]2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10Cl2Pd2
CAS Number:
Molecular Weight:
365.89
Beilstein:
4124623
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

reaction suitability

core: palladium
reaction type: Cross Couplings
reagent type: catalyst
reaction type: C-H Activation

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

SMILES string

Cl[Pd]CC=C.Cl[Pd]CC=C

InChI

1S/2C3H5.2ClH.2Pd/c2*1-3-2;;;;/h2*3H,1-2H2;2*1H;;/q;;;;2*+1/p-2

InChI key

TWKVUTXHANJYGH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Allylpalladium(II) chloride dimer is employed as catalyst in Heck reaction. It also participates as catalyst in the tandem nucleophilic allylation-alkoxyallylation reaction of the alkynylaldehydes with allyl chloride and allyltributylstannane.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Allylpalladium(II) chloride dimer has been employed for the following studies:
  • Synthesis of cationic palladium cataysts, used in the microwave-assisted Heck arylation.
  • Synthesis of N-heterocyclic carbene-palladium-η3-allyl chloride complexes, which are efficient catalyst for the Suzuki-Miyaura cross-coupling of aryl bromides and activated aryl chlorides.
  • Synthesis of 1,4-diallyl-1,2-dihydroisoquinolines.
  • As catalyst for greener Buchwald-Hartwig coupling in TPGS-750-M.

On the Way Towards Greener Transition-Metal-Catalyzed Processes as Quantified by E Factors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suzuki cross-couplings of alkyl tosylates that possess beta hydrogen atoms: synthetic and mechanistic studies.
Matthew R Netherton et al.
Angewandte Chemie (International ed. in English), 41(20), 3910-3912 (2002-10-19)
Efficient Heck vinylation of aryl halides catalyzed by a new air-stable palladium-tetraphosphine complex.
M Feuerstein et al.
The Journal of organic chemistry, 66(17), 5923-5925 (2001-08-21)
Tandem nucleophilic allylation-alkoxyallylation of alkynylaldehydes via amphiphilic bis-p-allylpalladium complexes.
Nakamura H, et al.
Tetrahedron Letters, 43(42), 7631-7633 (2002)
Radetich, B.; RajanBabu, T. V.
Journal of the American Chemical Society, 120, 8007-8007 (1998)
An N-heterocyclic carbene-palladium-η3-allyl chloride complex for the Suzuki-Miyaura coupling of aryl halides.
Broekemier NW, et al.
European Journal of Chemistry, 5(1), 162-166 (2014)

Articles

A variety of palladium-catalyzed cross-coupling reactions can be run under room temperature conditions in water with TPGS- 750-M, using a variety of commercially available palladium complexes and ligands.

A variety of palladium-catalyzed cross-coupling reactions can be run under room temperature conditions in water with TPGS- 750-M, using a variety of commercially available palladium complexes and ligands.

A variety of palladium-catalyzed cross-coupling reactions can be run under room temperature conditions in water with TPGS- 750-M, using a variety of commercially available palladium complexes and ligands.

A variety of palladium-catalyzed cross-coupling reactions can be run under room temperature conditions in water with TPGS- 750-M, using a variety of commercially available palladium complexes and ligands.

Protocols

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service