Skip to Content
Merck
All Photos(1)

Key Documents

697559

Sigma-Aldrich

Lithium-6Li hexafluorophosphate

95 atom % 6Li, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
6LiPF6
CAS Number:
Molecular Weight:
150.98
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

95 atom % 6Li

Assay

98% (CP)

form

solid

mp

200 °C (dec) (lit.)

mass shift

M-1

SMILES string

[6Li+].F[P-](F)(F)(F)(F)F

InChI

1S/F6P.Li/c1-7(2,3,4,5)6;/q-1;+1/i;1-1

InChI key

AXPLOJNSKRXQPA-HCMAANCNSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service