Skip to Content
Merck
All Photos(1)

Key Documents

601497

Sigma-Aldrich

Lithium-7Li fluoride

99 atom % 7Li, 99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
7LiF
CAS Number:
Molecular Weight:
26.01
EC Number:
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:

isotopic purity

99 atom % 7Li

Assay

99% (CP)

form

solid

mass shift

depleted

SMILES string

[7Li+].[F-]

InChI

1S/FH.Li/h1H;/q;+1/p-1/i;1+0

InChI key

PQXKHYXIUOZZFA-QMVMUTFZSA-M

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengyou Zhu et al.
Journal of hazardous materials, 399, 123054-123054 (2020-06-12)
Rational and smart design of hydrogen (H2) sensors especially those featured with flexibility and light weight is highly desirable, to meet the requirements for future development of portable H2 sensors. In this work, we demonstrate a novel paper film H2
Kan Usuda et al.
Biological trace element research, 167(1), 84-90 (2015-03-10)
Various fluoride compounds are widely used in industry. The present risk assessment study was conducted using a series of inorganic binary fluorides of the type XFn, where X(n) = Na(+), K(+), Li(+), Mg(2+), Ca(2+), Sr(2+), Ba(2+), Al(3+), Nd(3+), La(3+), Ce(3+), Sm(3+), Gd(3+)
Brian J Worfolk et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(46), 14138-14143 (2015-10-31)
With consumer electronics transitioning toward flexible products, there is a growing need for high-performance, mechanically robust, and inexpensive transparent conductors (TCs) for optoelectronic device integration. Herein, we report the scalable fabrication of highly conductive poly(3,4-ethylenedioxythiophene):poly(styrenesulfonate) ( PSS) thin films via

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service