Skip to Content
Merck
All Photos(3)

Key Documents

340421

Sigma-Aldrich

Lithium-6Li

chunks, in mineral oil, 95 atom % 6Li

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
6Li
CAS Number:
Molecular Weight:
6.02
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

isotopic purity

95 atom % 6Li

form

chunks

mass shift

M-1

SMILES string

[6Li]

InChI

1S/Li/i1-1

InChI key

WHXSMMKQMYFTQS-BJUDXGSMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium-6Li is an isolated and enriched isotope of lithium that is majorly used in fusion reactors. It is an important element in isotope separation study and can be sperated in large scale by using amalgam method. It is incorpoared as a blanket material for deuterium tritium fusion power reactor.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium isotope effects accompanying electrochemical insertion of lithium into metal oxides
Mouri M, et al.
Journal of Nuclear Science and Technology (Abingdon, United Kingdom), 44(1), 73-80 (2007)
Journal of the American Chemical Society, 110, 3668-3668 (1988)
Chunyapuk Kukusamude et al.
Food chemistry, 334, 127402-127402 (2020-07-28)
In this study, heavy metals including Cr, Mn, Co, Ni, Cu, Zn, As, and Cd in 55 Thai local rice (4 varieties) were measured using ICP-MS. Health risks were estimated from various Thai population groups, classified according to ages and
Lithium isotope effects upon electrochemical release from lithium manganese oxide
Okano K, et al.
Energy Procedia, 71(1), 140-148 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service