Skip to Content
Merck
All Photos(1)

Key Documents

695149

Sigma-Aldrich

DTBPF

Synonym(s):

1,1′-Bis(di-tert-butylphosphino)ferrocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44FeP2
CAS Number:
Molecular Weight:
474.42
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Arylations

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Indole Forming Reactions

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

mp

73-75 °C

functional group

phosphine

SMILES string

[Fe].CC(C)(C)P([C]1[CH][CH][CH][CH]1)C(C)(C)C.CC(C)(C)P([C]2[CH][CH][CH][CH]2)C(C)(C)C

InChI

1S/2C13H22P.Fe/c2*1-12(2,3)14(13(4,5)6)11-9-7-8-10-11;/h2*7-10H,1-6H3;

InChI key

FPLSJBJGQLJLSV-UHFFFAOYSA-N

Application

Form the diene complexation; carbonyl insertion; cycloaddition; reduction and deoxygenation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darbasie, N. D.; Schnatter, W. F. K. et al.
Tetrahedron Letters, 47, 963-963 (2004)
Sergeyev, S.; Hesse, M.
Synlett, 8, 1313-1313 (2002)
Anthony J Pearson et al.
Journal of the American Chemical Society, 125(44), 13326-13327 (2003-10-30)
A convenient one-pot procedure to prepare angularly substituted bicyclic and tricyclic molecules with excellent diastereoselectivity in good yield was developed, by Fe(CO)5 promoted cyclization. Three transformations (complexation, isomerization, and cyclization) were realized in a single operation. The product of this
Sean H Wiedemann et al.
The Journal of organic chemistry, 71(5), 1969-1976 (2006-02-25)
The inter- and intramolecular couplings of unactivated alkenes to 3,4-dihydroquinazolines with a Rh(I) catalyst are reported. Coupling between olefins and NH-3,4-dihydroquinazoline was found to occur consecutively with heterocycle dehydrogenation in the presence of a Rh(I)/PCy3/HCl catalyst. The reaction was used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service