Skip to Content
Merck
All Photos(5)

Key Documents

678740

Sigma-Aldrich

Bis(di-tert-butyl(4-dimethylaminophenyl)phosphine)dichloropalladium(II)

Synonym(s):

(A-taPhos)2PdCl2, Pd(amphos)Cl2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H56Cl2N2P2Pd
CAS Number:
Molecular Weight:
708.07
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

functional group

phosphine

SMILES string

Cl[Pd]Cl.CN(c1ccc(P(C(C)(C)C)C(C)(C)C)cc1)C.CN(c2ccc(P(C(C)(C)C)C(C)(C)C)cc2)C

InChI

1S/2C16H28NP.2ClH.Pd/c2*1-15(2,3)18(16(4,5)6)14-11-9-13(10-12-14)17(7)8;;;/h2*9-12H,1-8H3;2*1H;/q;;;;+2/p-2

InChI key

DWOZNANUEDYIOF-UHFFFAOYSA-L

General description

Bis(di-tert-butyl(4-dimethylaminophenyl)phosphine) dichloropalladium(II) is an organic compound with chemical formula C32H56Cl2N2P2Pd. It is commonly used as a catalyst in the organic synthesis.

For small scale and high throughput uses, product is also available as ChemBeads (927791)

Application

Bis(di-tert-butyl(4-dimethylaminophenyl)phosphine)dichloropalladium(II) is used as a catalyst:

  • In the enantioselective construction of indole-fused bicyclo[3.2.1]-octanes via an aminopalladition-triggered Heck-type reaction.
  • In the synthesis of phenanthridine derivatives from ortho bromo N-tosylhydrazones and 2-aminophenylboronic ester via Suzuki cross-coupling reaction followed by intramolecular condensation reaction.
  • In the synthesis of indenones by Pd-catalyzed annulation of an ortho-halobenzyl alcohol with internal alkynes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave assisted synthesis of phenanthridine derivatives via Suzuki coupling and condensation
Dende SK, et al.
Results in Chemistry, 3, 00149-00149 (2021)
Palladium-catalyzed annulation of alkynes with ortho-halide-containing benzyl alcohols in aqueous medium
Feng J, et al.
The Journal of Organic Chemistry, 79(21), 10561-10567 (2014)
Enantioselective Synthesis of Indole-Fused Bicyclo [3.2. 1] octanes via Palladium (II)-Catalyzed Cascade Reaction
Wang G, et al.
Organic Letters, 23(3) (2021)
Arkady Krasovskiy et al.
Journal of the American Chemical Society, 131(43), 15592-15593 (2009-10-16)
Mix in water, stir. That is all that is required in this new approach to sp(3)-sp(2) cross-couplings between an alkyl iodide and an aryl bromide, both potentially bearing functionality. They react under catalysis by Pd(0) in the presence of zinc
Anil S Guram et al.
Organic letters, 8(9), 1787-1789 (2006-04-21)
[reaction: see text] New air-stable PdCl(2){P(t)Bu(2)(p-R-Ph)}(2) (R = H, NMe(2), CF(3),) complexes represent simple, general, and efficient catalysts for the Suzuki-Miyaura cross-coupling reactions of aryl halides including five-membered heteroaryl halides and heteroatom-substituted six-membered heteroaryl chlorides with a diverse range of

Articles

Ligand used to prepare a palladium dichloride catalyst on treatment with PdCl2(COD). The catalyst effectively cross-couples aryl boronic acids with heteroaryl chlorides.

A variety of palladium-catalyzed cross-coupling reactions can be run under room temperature conditions in water with TPGS- 750-M, using a variety of commercially available palladium complexes and ligands.

A variety of palladium-catalyzed cross-coupling reactions can be run under room temperature conditions in water with TPGS- 750-M, using a variety of commercially available palladium complexes and ligands.

A variety of palladium-catalyzed cross-coupling reactions can be run under room temperature conditions in water with TPGS- 750-M, using a variety of commercially available palladium complexes and ligands.

See All

Protocols

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service