692050
4-(Trifluoromethyl)phenethyl bromide
97%
Synonym(s):
1-(2-Bromoethyl)-4-(trifluoromethyl)benzene, 2-[4-(Trifluoromethyl)phenyl]ethyl bromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H8BrF3
CAS Number:
Molecular Weight:
253.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n20/D 1.487
density
1.499 g/mL at 25 °C
functional group
bromo
fluoro
SMILES string
FC(F)(F)c1ccc(CCBr)cc1
InChI
1S/C9H8BrF3/c10-6-5-7-1-3-8(4-2-7)9(11,12)13/h1-4H,5-6H2
InChI key
WTCVMJLGKMOROW-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service