Skip to Content
Merck
All Photos(1)

Key Documents

370622

Sigma-Aldrich

4-(Trifluoromethoxy)benzyl bromide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4CH2Br
CAS Number:
Molecular Weight:
255.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.48 (lit.)

bp

82-84 °C/10 mmHg (lit.)

density

1.594 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

FC(F)(F)Oc1ccc(CBr)cc1

InChI

1S/C8H6BrF3O/c9-5-6-1-3-7(4-2-6)13-8(10,11)12/h1-4H,5H2

InChI key

JDNPUJCKXLOHOW-UHFFFAOYSA-N

General description

Polymerizations of 4-(trifluoromethoxy)benzyl bromide, via Friedel-Crafts polymerization using aluminum chloride as a catalyst has been reported.

Application

4-(Trifluoromethoxy)benzyl bromide may be used in the synthesis of bioreductive drug, (6S)-2-nitro-6-{[4-(trifluoromethoxy)benzyl]oxy}-6,7-dihydro-5H-imidazo[2,1-b][1,3]oxazine (PA-824).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew M Thompson et al.
Journal of medicinal chemistry, 52(3), 637-645 (2008-12-23)
The nitroimidazooxazine S-1 (PA-824) is a new class of bioreductive drug for tuberculosis. A series of related bicyclic nitroheterocycles was synthesized, designed to have a wide range of one-electron reduction potentials E(1) (from -570 to -338 mV, compared with -534
Synthesis and properties of fluorine-containing poly (arylenemethylene) s as new heat resistant denatured phenolic resins.
Nemoto T, et al.
Polymer Journal, 40(7), 622-628 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service