Skip to Content
Merck
All Photos(1)

Key Documents

686166

Sigma-Aldrich

2-Amino-5-methyl-1,3,4-thiadiazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5N3S
CAS Number:
Molecular Weight:
115.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

223-228 °C

SMILES string

Cc1nnc(N)s1

InChI

1S/C3H5N3S/c1-2-5-6-3(4)7-2/h1H3,(H2,4,6)

InChI key

HMPUHXCGUHDVBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jingjing Zhang et al.
International journal of biological macromolecules, 141, 1271-1279 (2019-09-08)
Several starch derivatives bearing urea groups (TUCAST, MTUCAST, and PTUCAST) based on 6-O-chloroacetylated starch (CAST) were successfully designed and synthesized. FT-IR, 1H NMR spectroscopy, and elemental analysis were carried out to identify the structural characteristics of starch derivatives. The antioxidant
Jingjing Zhang et al.
Carbohydrate polymers, 215, 108-118 (2019-04-15)
In the current study, five novel urea-functionalized chitosan derivatives were synthesized via condensation reactions of chloroacetyl chitosan (CTCS) with urea groups bearing nitrogen-containing heterocycles. In order to identify the structure characteristics of chitosan derivatives, FT-IR, 1H NMR spectroscopy, and elemental
Jingjing Zhang et al.
Carbohydrate polymers, 234, 115903-115903 (2020-02-20)
In this study, 2-urea-chitosan oligosaccharide derivatives (2-urea-COS derivatives) and 2,6-diurea-chitosan oligosaccharide derivatives (2,6-diurea-COS derivatives) were successfully designed and synthesized via intermediate 2-methoxyformylated chitosan oligosaccharide. All samples were characterized and compared based on FT-IR, 1H NMR spectroscopy, and elemental analysis. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service