Skip to Content
Merck
All Photos(1)

Key Documents

494216

Sigma-Aldrich

2-Amino-5-(methylthio)-1,3,4-thiadiazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5N3S2
CAS Number:
Molecular Weight:
147.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

178-181 °C (lit.)

SMILES string

CSc1nnc(N)s1

InChI

1S/C3H5N3S2/c1-7-3-6-5-2(4)8-3/h1H3,(H2,4,5)

InChI key

PCLAZAJARAIGGD-UHFFFAOYSA-N

General description

2-Amino-5-(methylthio)-1,3,4-thiadiazole is a 2-amino-5-substituted-1,3,4-thiadiazole. Its density at 25°C is 1.4020g/ml.

Application

2-Amino-5-(methylthio)-1,3,4-thiadiazole may be used in the synthesis of 9-methylthio-7,7-dioxo-7,7a-dihydro-5-oxo-7λ6,10-dithia-8,11-diaza-cyclopenta[b] phenantren-6-one by reacting with 4-chloro-2-oxo-2H-chromene-3-sulfonyl chloride.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Microbiological Activity of Some Newly Synthesized Derivatives of 2-Oxo-2H-chromen-2-one.
Daci-Ajvazi M, et al.
Journal of Chemistry, 8(4), 1522-1527 (2011)
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 268-268 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service