Skip to Content
Merck
All Photos(3)

Key Documents

683213

Sigma-Aldrich

Chiralyst P612

Umicore, 97%

Synonym(s):

Dichloro(p-cymene)ruthenium(II) dimer, Di(μ-chloro)dichlorobis(p-cymene)diruthenium(II), (p-Cymene)ruthenium(II) chloride dimer, Benzene, 1-methyl-4-(1-methylethyl)-, ruthenium complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Ru(p-cymene)Cl2]2
CAS Number:
Molecular Weight:
612.39
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

reaction suitability

core: ruthenium
reaction type: Cross Couplings
reagent type: catalyst
reaction type: C-H Activation

mp

247.0-250.0 °C (dec.) (lit.)

SMILES string

Cl[Ru]Cl.Cl[Ru]Cl.CC(C)c1ccc(C)cc1.CC(C)c2ccc(C)cc2

InChI

1S/2C10H14.4ClH.2Ru/c2*1-8(2)10-6-4-9(3)5-7-10;;;;;;/h2*4-8H,1-3H3;4*1H;;/q;;;;;;2*+2/p-4

InChI key

LAXRNWSASWOFOT-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Cyclometalated ruthenium complexes with arylimines and nitrogen-containing heterocycles via C-H bond activation

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Li et al.
Dalton transactions (Cambridge, England : 2003), 41(36), 10934-10937 (2012-08-15)
The reaction of [RuCl(2)(p-cymene)](2) with arylimines and 4 equiv. of KOAc in methanol at room temperature produces stable (N^C)-cyclometallated ruthenium(II) complexes via C-H bond activation/deprotonation. This method can be applied also to nitrogen-containing molecules: N-phenylpyrazole, 2-phenyl-2-oxazoline and benzo[h]quinoline. N-Phenyl-pyrazole, [RuCl(2)(p-cymene)](2)

Articles

Chiral vicinal diamines are of tremendous interest to the synthetic chemist as they are found in many chiral catalysts and pharmaceuticals.

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service