![Chiralyst P612 Umicore, 97%](/deepweb/assets/sigmaaldrich/product/structures/226/369/c9921f4d-5a66-49a7-b1a8-3ab3fe3d0086/640/c9921f4d-5a66-49a7-b1a8-3ab3fe3d0086.png)
Chiralyst P612
![Chiralyst P612 Umicore, 97%](/deepweb/assets/sigmaaldrich/product/structures/226/369/c9921f4d-5a66-49a7-b1a8-3ab3fe3d0086/640/c9921f4d-5a66-49a7-b1a8-3ab3fe3d0086.png)
Synonym(s):
Dichloro(p-cymene)ruthenium(II) dimer, (p-Cymene)ruthenium(II) chloride dimer, Benzene, 1-methyl-4-(1-methylethyl)-, ruthenium complex
Linear Formula:
[Ru(p-cymene)Cl2]2
CAS Number:
Molecular Weight:
612.39
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing