Skip to Content
Merck
All Photos(2)

Key Documents

684147

Sigma-Aldrich

(1S, 2S)-1,2-di-1-Naphthyl-ethylenediamine dihydrochloride

97%

Synonym(s):

(S, S)-1,2-Bis(1-naphthyl)-1,2-ethanediamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20N2 · 2HCl
CAS Number:
Molecular Weight:
385.33
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

optical activity

[α]22/D +258.0°, c = 1 in H2O

mp

219-224 °C

SMILES string

Cl[H].Cl[H].N[C@H]([C@@H](N)c1cccc2ccccc12)c3cccc4ccccc34

InChI

1S/C22H20N2.2ClH/c23-21(19-13-5-9-15-7-1-3-11-17(15)19)22(24)20-14-6-10-16-8-2-4-12-18(16)20;;/h1-14,21-22H,23-24H2;2*1H/t21-,22-;;/m0../s1

InChI key

SHNGCXWOHADIKG-IXOXMDGESA-N

Application

(1S, 2S)-1,2-di-1-Naphthyl-ethylenediamine dihydrochloride can be used to synthesize N,N′-[(1S,2S)-1,2-di-1-naphthyl-ethylene]bis2-propenamide, a chiral monomer, to prepare polymeric chiral stationary phase for HPLC.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tharanga Payagala et al.
Analytical and bioanalytical chemistry, 399(7), 2445-2461 (2011-01-05)
Three new polymeric chiral stationary phases were synthesized based on (1S,2S)-1,2-bis(2,4,6-trimethylphenyl)ethylenediamine, (1S,2S)-1,2-bis(2-chlorophenyl)ethylenediamine, and (1S,2S)-1,2-di-1-naphthylethylenediamine via a simple free-radical-initiated polymerization in solution. These monomers are structurally related to (1S,2S)-1,2-diphenylethylenediamine which is the chiral monomer used for the commercial P-CAP-DP polymeric chiral

Articles

Chiral vicinal diamines are of tremendous interest to the synthetic chemist as they are found in many chiral catalysts and pharmaceuticals.

Related Content

The Chin group is interested in computational and experimental approaches to understanding stereoselective recognition and catalysis. Their studies in weak forces (H-bonding, electronic and steric effects) has led to a highly efficient method for making limitless varieties of chiral vicinal diamines from the 'mother diamine' that are useful for developing stereoselective organocatalysts or transition metal-based catalysts as well as for developing drugs (Acc Chem Res (2012) p1345). The 'mother diamine' is also useful for making binol, monophos and binap analogs. The Chin group is also interested in using reversible covalent bonds for stereoselective recognition and L to D conversion of natural and non-natural amino acids (EJOC (2012) p229).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service