Skip to Content
Merck
All Photos(1)

Key Documents

671460

Sigma-Aldrich

Propanoic acid hydrazide

≥90% (GC)

Synonym(s):

Propanoic acid hydrazide, Propanoic hydrazide, Propionylhydrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H8N2O
CAS Number:
Molecular Weight:
88.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥90% (GC)

functional group

amine
hydrazine

SMILES string

CCC(=O)NN

InChI

1S/C3H8N2O/c1-2-3(6)5-4/h2,4H2,1H3,(H,5,6)

InChI key

DXGIRFAFSFKYCF-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yufan Zhang et al.
Journal of biomaterials science. Polymer edition, 30(12), 995-1007 (2019-05-16)
A series of injectable in situ dual-crosslinking hydrogels (HA/ALG) based on oxidized sodium alginate (oxi-ALG) and hyaluronic acid modified with thiol and hydrazide (HA-SH/CDH) were prepared via hydrazone bonds and disulfide bonds. The chemical structures, morphologies, rheological properties, gelling time
Arturo Matamoros-Volante et al.
Journal of cell science, 133(2) (2020-01-15)
Capacitation in mammalian sperm involves the accurate balance of intracellular pH (pHi), but the mechanisms controlling this process are not fully understood, particularly regarding the spatiotemporal regulation of the proteins involved in pHi modulation. Here, we employed an image-based flow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service