Skip to Content
Merck
All Photos(3)

Key Documents

109010

Sigma-Aldrich

3,3′-Dithiodipropionic acid

99%

Synonym(s):

2-Carboxyethyl disulfide, Bis(2-carboxyethyl) disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCOCH2CH2SSCH2CH2COOH
CAS Number:
Molecular Weight:
210.27
Beilstein:
1778543
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

155-158 °C (lit.)

functional group

carboxylic acid
disulfide

SMILES string

OC(=O)CCSSCCC(O)=O

InChI

1S/C6H10O4S2/c7-5(8)1-3-11-12-4-2-6(9)10/h1-4H2,(H,7,8)(H,9,10)

InChI key

YCLSOMLVSHPPFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,3′-Dithiodipropionic acid forms monolayers on a polycrystalline gold electrode through a self-assembly procedure to form gold 3,3′-dithiodipropionic acid self-assembled monolayer modified electrode. It is the precursor substrate in the synthesis of 3-mercaptopropionic acid-containing polythioesters. It is added as primary carbon supplement in the culture medium of novel betaproteobacterium, strain DPN7.

Application

3,3′-Dithiodipropionic acid was used as capping agent for introducing charge on gold nanoparticles surfaces.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuguang Zhang et al.
Advanced healthcare materials, 9(18), e2000387-e2000387 (2020-08-21)
It is of great significance to develop multifunctional biomaterials to effectively deliver anticancer drug to tumor cells for cancer therapy. Here, inspired by the specific tumor microenvironment (TME) cues, a unique multistage pH/redox-responsive polyprodrug composed of amphiphilic pH-sensitive diblock copolymer
Jian-Tao Lin et al.
Colloids and surfaces. B, Biointerfaces, 155, 41-50 (2017-04-14)
Stimuli-responsive nanocarriers for anticancer drug and gene co-delivery are promising strategy in cancer therapy. The ultimate goal is to deliver high local concentration of therapeutic agents with no premature release and result in synergistic effects for combination therapies. In this
Yuling Liu et al.
International journal of nanomedicine, 12, 2635-2644 (2017-04-25)
Pluronic F127 (F127), an amphiphilic triblock copolymer, has been shown to have significant potential for drug delivery, as it is able to incorporate hydrophobic drugs and self-assemble into nanosize micelles. However, it suffers from dissociation upon dilution owing to the
Xiaoyou Xu et al.
Journal of chromatography. A, 1167(1), 35-41 (2007-09-07)
We employed agarose gel preparative electrophoresis to separate gold nanoparticles based on size, shape, and charge. The separating technique was first demonstrated by size separation of 5 nm, 15 nm, and 20 nm spherical gold nanoclusters; and further evidenced through
Yanqin Xu et al.
Journal of biomaterials science. Polymer edition, 31(13), 1706-1721 (2020-07-03)
A pH and redox dual-responsive composite hydrogel was developed, which was formed from triblock copolymer PEG-SS-PCL-SS-PEG and Ag/AgO/carboxymethyl chitosan (CMCS) hydrogel inclusion complexes. Dithiodipropionic acid and poly(ethylene glycol) (PEG) were esterified to synthesize a hydrophilic segment PEG-SS-COOH polymer containing a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service