Skip to Content
Merck
All Photos(1)

Key Documents

651621

Sigma-Aldrich

4-Amino-3-nitrophenylboronic acid

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7BN2O4
CAS Number:
Molecular Weight:
181.94
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

form

solid

mp

215-225 °C (dec.)

SMILES string

Nc1ccc(cc1[N+]([O-])=O)B(O)O

InChI

1S/C6H7BN2O4/c8-5-2-1-4(7(10)11)3-6(5)9(12)13/h1-3,10-11H,8H2

InChI key

IRTXQNNJQZNKRP-UHFFFAOYSA-N

General description

4-Amino-3-nitrophenylboronic acid is a synthetic building block used in metal-catalyzed Suzuki coupling reaction.

Application

4-Amino-3-nitrophenylboronic acid can be used as a reactant to prepare difluoromethoxy-3-nitrobiphenyl-4-amine by coupling with 1-bromo-2-(difluoromethoxy)-benzene in the presence of a palladium catalyst.

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design and optimization of benzimidazole-containing transient receptor potential melastatin 8 (TRPM8) antagonists
Parks DJ, et al.
Journal of medicinal chemistry, 54(1), 233-247 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service