Skip to Content

End of year dates: Orders must be billed by 05/12. Merck Mexico will be closed from 18/12 to 02/01.

Merck

643513

Sigma-Aldrich

2-Amino-5-chloro-3-methylbenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H2(Cl)(CH3)CO2H
CAS Number:
Molecular Weight:
185.61
MDL number:
UNSPSC Code:
12352106
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

239-243 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cc1cc(Cl)cc(c1N)C(O)=O

InChI

1S/C8H8ClNO2/c1-4-2-5(9)3-6(7(4)10)8(11)12/h2-3H,10H2,1H3,(H,11,12)

InChI key

KOPXCQUAFDWYOE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service