Skip to Content
Merck
All Photos(2)

Key Documents

638730

Sigma-Aldrich

2-Chloro-3,4-dimethoxybenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H2(Cl)CO2H
CAS Number:
Molecular Weight:
216.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

200-204 °C (lit.)

SMILES string

COc1ccc(C(O)=O)c(Cl)c1OC

InChI

1S/C9H9ClO4/c1-13-6-4-3-5(9(11)12)7(10)8(6)14-2/h3-4H,1-2H3,(H,11,12)

InChI key

YSEIYOHXERRMNN-UHFFFAOYSA-N

Application

2-Chloro-3,4-dimethoxybenzoic acid can be used as a starting material in the four-step synthesis of 14C-labelled N-(2-chloro-3,4-dimethoxybenzylideneamino)guanidinium acetate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Synthesis of N-(2-chloro-3, 4-dimethoxybenzylideneamino) guanidinium acetate [a-14C]"
Almeida M, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 45(05), 371-377 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service