Skip to Content
Merck
All Photos(2)

Key Documents

196886

Sigma-Aldrich

2-(Trifluoromethyl)benzoic acid

98%

Synonym(s):

α,α,α-Trifluoro-o-toluic acid, 2-Carboxybenzotrifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CO2H
CAS Number:
Molecular Weight:
190.12
Beilstein:
976984
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

247 °C/753 mmHg (lit.)

mp

107-110 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)c1ccccc1C(F)(F)F

InChI

1S/C8H5F3O2/c9-8(10,11)6-4-2-1-3-5(6)7(12)13/h1-4H,(H,12,13)

InChI key

FBRJYBGLCHWYOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2-(Trifluoromethyl)benzoic acid was used in the synthesis of 1,3,4-oxadiazole derivatives containing 2-fluoro-4-methoxy moiety. It was also used to investigate the binding of 2-pyridinone and amino acid derivative as ligands with chaperones PapD and FimC by surface plasmon resonance and 1HNMR spectroscopy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R D Farrant et al.
Journal of pharmaceutical and biomedical analysis, 13(8), 971-977 (1995-07-01)
Many drugs containing carboxylic acid functional groups are metabolised in vivo to ester glucuronides (1-O-acyl-beta-D-glucopyranuronates) and, of these, a number show a propensity to undergo internal isomerisation via a transacylation process, causing the carboxylic acid moiety to migrate successively to
Tobias Tengel et al.
Organic & biomolecular chemistry, 2(5), 725-731 (2004-02-27)
Identification of compounds from chemical libraries that bind to macromolecules by use of NMR spectroscopy has gained increasing importance during recent years. A simple methodology based on (19)F NMR spectroscopy for the screening of ligands that bind to proteins, which
B Chandrakantha et al.
European journal of medicinal chemistry, 45(3), 1206-1210 (2009-12-17)
In the present study a series of new 1,3,4-oxadiazole derivatives containing 2-fluoro-4-methoxy moiety were synthesized. These newly synthesized compounds were characterized by NMR, mass spectral, IR spectral study and also by C, H, N analyses. All the newly synthesized compounds
Studies on the metabolism of fluorinated xenobiotics in the rat using 19F-NMR and 1H-NMR spectroscopy.
F Y Ghauri et al.
Journal of pharmaceutical and biomedical analysis, 8(8-12), 939-944 (1990-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service