Skip to Content
Merck
All Photos(1)

Key Documents

632058

Sigma-Aldrich

4-Phenyl-1-butyne

97%

Synonym(s):

(3-Butynyl)benzene, 1-Phenyl-3-butyne, 3-Butyn-1-ylbenzene, Phenethylacetylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C2H4C≡CH
CAS Number:
Molecular Weight:
130.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5120 (lit.)

bp

189-190 °C (lit.)

density

0.926 g/mL at 25 °C (lit.)

functional group

phenyl

SMILES string

C#CCCc1ccccc1

InChI

1S/C10H10/c1-2-3-7-10-8-5-4-6-9-10/h1,4-6,8-9H,3,7H2

InChI key

QDEOKXOYHYUKMS-UHFFFAOYSA-N

Application

4-Phenyl-1-butyne can undergo addition reaction with α-imino esters to give the corresponding β,γ-alkynyl α-amino acid derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Efficient Synthesis of ?, ?-Alkynyl a-Amino Acid Derivatives by Ag (I)-Catalyzed Alkynylation of a-Imino Esters"
Ji X-J, et al.
Advanced Synthesis & Catalysis, 346(1), 42- 44 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service