Skip to Content
Merck
All Photos(1)

Key Documents

206504

Sigma-Aldrich

4-Ethynyltoluene

97%

Synonym(s):

1-Ethynyl-4-methylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4C≡CH
CAS Number:
Molecular Weight:
116.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.547 (lit.)

bp

168-170 °C (lit.)

density

0.916 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(cc1)C#C

InChI

1S/C9H8/c1-3-9-6-4-8(2)5-7-9/h1,4-7H,2H3

InChI key

KSZVOXHGCKKOLL-UHFFFAOYSA-N

Application

4-Ethynyltoluene was used in the synthesis of bis(bicyclic) products by the cycloaddition of alkynes to the AlN2C2[1].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanxia Zhao et al.
Chemical communications (Cambridge, England), 49(40), 4546-4548 (2013-04-09)
The reaction of dialumane, [L(2-)(THF)Al(II)-Al(II)(THF)L(2-)] (1, L = [(2,6-iPr2C6H3)NC(Me)]2), with the less reactive diphenylacetylene affords a new insertion product [L(2-)(THF)Al(III)(PhC=CPh)(2-)Al(III)(THF)L(2-)] (2). Reaction of 1 with phenylacetylene (or 4-ethynyltoluene), however, proceeds through cycloaddition of alkynes to the AlN2C2 rings to produce
Li-Jing Han et al.
Dalton transactions (Cambridge, England : 2003), 49(24), 8347-8353 (2020-06-11)
A series of dinuclear platinum(ii) alkynyl complexes [Pt2L2(C[triple bond, length as m-dash]CC6H4R-4)4] (R = H 1, CH32, But3) and unusual tetranuclear Pt(ii)-Ag(i) clusters [Pt2Ag2L(C[triple bond, length as m-dash]CC6H4R-4)6] (R = H, 4; CH3, 5; But, 6), together with novel polymer
Jih Ru Hwu et al.
Molecules (Basel, Switzerland), 25(17) (2020-08-28)
The modern world has no available drugs for the treatment of enteroviruses (EV), which affect millions of people worldwide each year. The EV71 is a major causative disease for hand, foot, and mouth disease; sometimes it is associated with severe
Amit N Pandya et al.
Tetrahedron letters, 55(50), 6922-6924 (2014-12-17)
An efficient strategy for the synthesis of indolizines from readily available starting materials via oxidative C-H functionalization and
Hanfeng Jin et al.
Rapid communications in mass spectrometry : RCM, 34(21), e8899-e8899 (2020-07-18)
Aromatic hydrocarbons play an important role in the formation and growth of polycyclic aromatic hydrocarbon (PAH) and soot particles. Measurements of their absolute photoionization cross-sections (PICSs), that benefit the quantitative investigation of mass spectrometry, are still lacking, however. PICSs of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service