Skip to Content
Merck
All Photos(2)

Key Documents

161241

Sigma-Aldrich

1-Phenyl-1-propyne

99%

Synonym(s):

(2-Methylethynyl)benzene, 1-Methyl-2-phenylacetylene, 1-Propyn-1-ylbenzene, 1-Propynylbenzene, 3-Phenyl-2-propyne, Methylphenylacetylene, Methylphenylethyne, Phenylmethylacetylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C≡CCH3
CAS Number:
Molecular Weight:
116.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.564 (lit.)

bp

185 °C (lit.)

density

0.928 g/mL at 25 °C (lit.)

functional group

phenyl

storage temp.

2-8°C

SMILES string

CC#Cc1ccccc1

InChI

1S/C9H8/c1-2-6-9-7-4-3-5-8-9/h3-5,7-8H,1H3

InChI key

GHUURDQYRGVEHX-UHFFFAOYSA-N

General description

Co-catalyzed reaction between cyclopentene and 1-phenyl-1-propyne has been reported. Bonding properties of 1-phenyl-1-propyne on Cu(111) at 100K have been studied using temperature-programmed desorption and X-ray, ultraviolet and two-photon photoemission spectroscopies. 1-Phenyl-1-propyne is an inhibitor of dopamine beta-hydroxylase. Polymerization of 1-phenyl-1-propyne by TaCl5 and NbCl5 has been reported.

1-Phenyl-1-propyne serves as a starting material for various chemical reactions, such as addition reactions and polymerization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(Arylimido)niobium(V)?Alkylidenes, Nb(CHSiMe3)(NAr)[OC(CF3)3](PMe3)2, That Enable to Proceed Living Metathesis Polymerization of Internal Alkynes
Izawa T and Nomura K
Macromolecules, 53(13), 5266-5279 (2020)
Youngku Sohn et al.
Langmuir : the ACS journal of surfaces and colloids, 23(24), 12185-12191 (2007-10-31)
The bonding properties of 1-phenyl-1-propyne (PP, C6H5CCCH3) on Cu(111) at 100 K have been studied using temperature-programmed desorption (TPD), and X-ray, ultraviolet, and two-photon photoemission spectroscopies (XPS, UPS, and 2PPE). In TPD, there is no evidence for dissociation. Multilayer desorption
Shuhuai Xiang et al.
Journal of the American Chemical Society, 136(16), 5832-5835 (2014-04-12)
Here we report a general method for the measurement of (13)C kinetic isotope effects at natural abundance for reactions that yield two or more products concurrently. We use, as an example, a recently reported Co-catalyzed reaction between cyclopentene and 1-phenyl-1-propyne.
Effect of organometallic cocatalysts on the polymerization of 1-phenyl-1-propyne by tantalum pentachloride (TaCl5) and niobium pentachloride (NbCl5).
Masuda T, et al.
Macromolecules, 18(11), 2109-2113 (1985)
G Colombo et al.
The Journal of biological chemistry, 259(24), 15017-15020 (1984-12-25)
The catalytic action of dopamine beta-hydroxylase on 1-phenyl-1-propyne results in concomitant loss of enzyme activity. At pH 5.5 and 25 degrees C, 1-phenyl-1-propyne inactivates dopamine beta-hydroxylase in a mechanism-based fashion. The inactivation rate is first-order, follows saturation kinetics, and is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service