Skip to Content
Merck
All Photos(1)

Key Documents

631507

Sigma-Aldrich

1,10-Phenanthrolin-5-amine

97%

Synonym(s):

(1,10)-Phenanthrolin-5-ylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9N3
CAS Number:
Molecular Weight:
195.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

254-258 °C (lit.)

SMILES string

Nc1cc2cccnc2c3ncccc13

InChI

1S/C12H9N3/c13-10-7-8-3-1-5-14-11(8)12-9(10)4-2-6-15-12/h1-7H,13H2

InChI key

DKPSSMOJHLISJI-UHFFFAOYSA-N

Application

1,10-Phenanthrolin-5-amine can be used in the synthesis of bis-[1,10]phenanthrolin-5-yl-pyromellitic diimide (Bphen-PMD), which can find applications in organic light-emitting devices.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siew San Tan et al.
Physical chemistry chemical physics : PCCP, 19(37), 25734-25745 (2017-09-16)
An absorbance-based sensor employing ruthenium bipyridyl with a phenanthroline-fused benzoylthiourea moiety formulated as [Ru(ii)(bpy)
Mona Amiri et al.
Dalton transactions (Cambridge, England : 2003), 49(29), 10173-10184 (2020-07-16)
1,10-Phenanthroline (phen) was grafted to either indium tin oxide (ITO), fluorine-doped tin oxide (FTO), or titanium dioxide (TiO2) semiconductors (SC's) by electrochemical reduction of 5-diazo-phen. The phen ligand is bonded to the semiconductor at C5, and it can be handled
"Phenanthroline diimide as an organic electron-injecting material for organic light-emitting devices"
Lee H, et al.
Royal Society of Chemistry Advances, 2(23), 8762- 8767 (2012)
Kollur Shiva Prasad et al.
Molecules (Basel, Switzerland), 25(12) (2020-06-26)
Herein we report the synthesis and structural elucidation of two novel imine-based ligands, 2-(1,10-phenanthrolin-5-yl)imino)methyl)-5-bromophenol (PIB) and N-(1,10-phenanthrolin-5-yl)-1-(thiophen-3-yl)methanimine (PTM) ligands. An in vitro cytotoxicity assay of the synthesized molecules was carried out against breast, cervical, colorectal, and prostate cancer cell lines
Barbara Feist et al.
Mikrochimica acta, 186(2), 91-91 (2019-01-12)
Graphene oxide (GO) was chemically functionalized with 5-amino-1,10-phenanthroline. The resulting conjugate (phen-GO) was characterized by scanning electron microscopy and X-ray photoelectron spectroscopy. The experiments show that phen-GO has a high affinity for extraction of Pb(II) ions. Isotherms and kinetics fit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service