Skip to Content
Merck
All Photos(1)

Key Documents

305529

Sigma-Aldrich

5-Amino-8-hydroxyquinoline dihydrochloride

95%

Synonym(s):

5-Amino-8-quinolinol dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2O · 2HCl
CAS Number:
Molecular Weight:
233.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

solid

mp

279 °C (dec.) (lit.)

SMILES string

Cl[H].Cl[H].Nc1ccc(O)c2ncccc12

InChI

1S/C9H8N2O.2ClH/c10-7-3-4-8(12)9-6(7)2-1-5-11-9;;/h1-5,12H,10H2;2*1H

InChI key

VTQDJAUGGZFPOI-UHFFFAOYSA-N

Application

5-Amino-8-hydroxyquinoline dihydrochloride was used in synthesis of:
  • 5-acetamido-8-hydroxyquinoline hydrochloride[1]
  • 5-(p-tolylsulfonylimino)quinolin-8-one[2]
  • 5,8-quinolinesemiquinone via ferric chloride oxidation[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Soedjak et al.
Biochimica et biophysica acta, 1335(1-2), 73-90 (1997-04-17)
Streptonigrin semiquinone (SQ.), a free radical intermediate implicated in the biological functioning of the antitumor antibiotic streptonigrin has been prepared and its structural properties in solution have been characterized. Through the use of electron paramagnetic resonance spectroscopy, the spin densities
Sanford A Asher et al.
Analytical chemistry, 75(7), 1676-1683 (2003-04-23)
We developed a polymerized crystalline colloidal array photonic material that senses metal cations in water at low concentrations (PCCACS). Metal cations such as Cu2+, Co2+, Ni2+, and Zn2+ bind to 8-hydroxyquinoline groups covalently attached to the PCCACS. At low metal
Quinone Imines with a Fused Azine Ring: I. Synthesis and Hydrochlorination of 5-(p-Tolylsulfonylimino) quinolin-8-one.
Belov AV and Nichvoloda VM.
Russ. J. Org. Chem., 40(1), 93-96 (2004)
Thangaraj Anand et al.
Analytica chimica acta, 853, 596-601 (2014-12-04)
We have synthesized a new probe 5-((anthracen-9-ylmethylene) amino)quinolin-10-ol (ANQ) based on anthracene platform. The probe was tested for its sensing behavior toward heavy metal ions Hg(2+), Pb(2+), light metal Al(3+) ion, alkali, alkaline earth, and transition metal ions by UV-visible

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service